Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrahydropyran-4-acetic acid 97%

Supplier: AMBEED, INC
Danger

A191595-1G A191595-5G A191595-1G A191595-5G A191595-100MG A191595-10G A191595-250MG
103816-386EA 11 USD InStock
103816-386 103846-988 77084-832 77084-834 77111-786 77111-788 77111-790
Tetrahydropyran-4-acetic acid 97%
Tetrahydropyran-4-acetic acid
Precaución: for Research Use Only


Formula: C₇H₁₂O₃
MW: 144.1700 g/mol
Boiling Pt: 285.1 °C (760 mm Hg)
Melting Pt: 43…47 °C
Flash Pt: 119.2 °C
Storage Temperature: Ambient
MDL Number: MFCD01631204
CAS Number: 85064-61-5
UN: 1759
ADR: 8 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty