Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tetrakis(4-bromophenyl)methane 95%

Supplier: AMBEED, INC
Warning

A141102-1G A141102-100MG A141102-250MG A141102-1G A141102-5G
103802-952EA 46 USD InStock
103802-952 76984-196 76984-198 77299-202 77299-204
Tetrakis(4-bromophenyl)methane 95%
Tetrakis(4-bromophenyl)methane
Precaución: for Research Use Only


Formula: C₂₅H₁₆Br₄
MW: 636.02 g/mol
Boiling Pt: 603.8 °C (760 mm Hg)
Melting Pt: 312.5…313 °C
Flash Pt: 305.4 °C
Storage Temperature: Ambient
MDL Number: MFCD19688472
CAS Number: 105309-59-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty