Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Thiazolecarboxylic acid 98%

Supplier: AMBEED, INC
Warning

A409029-1G A409029-100G A409029-10G A409029-25G A409029-5G A409029-1G
103827-196EA 7 USD InStock
103827-196 103838-244 76773-784 76773-786 76841-758 76841-760 77307-132
4-Thiazolecarboxylic acid 98%
4-Thiazolecarboxylic acid
Precaución: for Research Use Only


Formula: C₄H₃NO₂S
MW: 129.1371 g/mol
Boiling Pt: 319.2 °C (760 mm Hg)
Melting Pt: 195…199 °C
Flash Pt: 146.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00623589
CAS Number: 3973-08-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty