Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thiazole-5-boronic acid pinacol ester 95%

Supplier: AMBEED, INC
Warning

A145105-1G A145105-5G A145105-250MG A145105-1G
103803-494EA 21 USD InStock
103803-494 103841-574 76972-790 77316-844
Thiazole-5-boronic acid pinacol ester 95%
Thiazole-5-boronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₉H₁₄BNO₂S
Melting Pt: 98…101 °C
Storage Temperature: Freezer
MDL Number: MFCD10697440
CAS Number: 1086111-09-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty