Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,3-Thiazolan-2-one 97%

Supplier: AMBEED, INC
Warning

A340119-5G A340119-5G A340119-100G A340119-10G A340119-25G
103836-780EA 5 USD InStock
103836-780 76839-794 77195-128 77195-130 77306-524
1,3-Thiazolan-2-one 97%
1,3-Thiazolan-2-one
Precaución: for Research Use Only


Formula: C₃H₅NOS
Boiling Pt: 247.6 °C (760 mm Hg)
Melting Pt: 53…54 °C
Flash Pt: 103.5 °C
Storage Temperature: Ambient
MDL Number: MFCD01332388
CAS Number: 2682-49-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty