Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Thiopheneboronic acid 98%

Supplier: AMBEED, INC
Warning

A240460-5G A240460-25G A240460-5G A240460-100G A240460-10G
103851-148EA 10 USD InStock
103851-148 77090-298 77090-300 77288-714 77288-716
2-Thiopheneboronic acid 98%
2-Thiopheneboronic acid
Precaución: for Research Use Only


Formula: C₄H₅BO₂S
MW: 127.96 g/mol
Boiling Pt: 287.9 °C (760 mm Hg)
Melting Pt: 138…140 °C
Flash Pt: 127.9 °C
Storage Temperature: Freezer
MDL Number: MFCD00151850
CAS Number: 6165-68-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty