Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Thiophenemethanol 97%

Supplier: AMBEED, INC
Warning

A131280-1G A131280-100G A131280-10G A131280-1G A131280-25G A131280-5G
103801-552EA 9 USD InStock
103801-552 77009-958 77009-960 77314-036 77314-038 77314-040
3-Thiophenemethanol 97%
3-Thiophenemethanol
Precaución: for Research Use Only


Formula: C₅H₆OS
Boiling Pt: 86…88 °C (760 mm Hg)
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00014534
CAS Number: 71637-34-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty