Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nɑ-Tosyl-L-arginine 98%

Supplier: AMBEED, INC
Warning

A367773-1G A367773-5G A367773-25G A367773-10G A367773-1G
103826-432EA 6 USD InStock
103826-432 103837-442 76840-684 77003-486 77306-792
Nɑ-Tosyl-L-arginine 98%
Nalpha-Tosyl-L-arginine
Precaución: for Research Use Only


Formula: C₁₃H₂₀N₄O₄S
Boiling Pt: 591.3 °C (760 mm Hg)
Melting Pt: 81…103 °C
Flash Pt: 311.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00019732
CAS Number: 1159-15-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty