Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-(4-Methylphenylsulfonamido)-3-phenylpropanoic acid ≥95%

Supplier: AMBEED, INC
Warning

A193757-5G A193757-10G A193757-100G
103847-192EA 11 USD InStock
103847-192 77667-612 77879-229
(S)-2-(4-Methylphenylsulfonamido)-3-phenylpropanoic acid ≥95%
(S)-2-(4-Methylphenylsulfonamido)-3-phenylpropanoic acid
Precaución: Sealed in dry, at room temperature.


Formula: C₁₆H₁₇NO₄S
MW: 319.4 g/mol
Boiling Pt: 517.7 °C (760 mm Hg)
Melting Pt: 165 °C
Flash Pt: 266.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00037251
CAS Number: 13505-32-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty