Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

E-Phenylethenylboronic acid 95%

Supplier: AMBEED, INC
Warning

A359042-5G A359042-1G A359042-5G
103826-216EA 11 USD InStock
103826-216 103837-224 76840-386 76840-388
E-Phenylethenylboronic acid 95%
E-Phenylethenylboronic acid
Precaución: for Research Use Only


Formula: C₈H₉BO₂
MW: 147.97 g/mol
Boiling Pt: 315.9 °C (760 mm Hg)
Melting Pt: 146…156 °C
Flash Pt: 144.9 °C
Storage Temperature: Freezer
MDL Number: MFCD00963621
CAS Number: 6783-05-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty