Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-4-Aminocyclohexanecarboxylic acid 97%

Supplier: AMBEED, INC
Warning

A233604-1G A233604-5G A233604-250MG A233604-1G A233604-25G A233604-5G
103810-890EA 18 USD InStock
103810-890 103850-652 76976-232 77089-608 77089-610 77089-612
trans-4-Aminocyclohexanecarboxylic acid 97%
trans-4-Aminocyclohexanecarboxylic acid
Precaución: for Research Use Only


Formula: C₇H₁₃NO₂
MW: 143.19 g/mol
Boiling Pt: 280 °C (760 mm Hg)
Melting Pt: >320 °C
Flash Pt: 123.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00209953
CAS Number: 3685-25-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty