Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-4-Ethylcyclohexanecarboxylic acid 98%

Supplier: AMBEED, INC
Warning

A671606-5G A671606-5G A671606-10G
103840-500EA 6 USD InStock
103840-500 77308-088 77747-361
trans-4-Ethylcyclohexanecarboxylic acid 98%
trans-4-Ethylcyclohexanecarboxylic acid
Precaución: for Research Use Only


Formula: C₉H₁₆O₂
Boiling Pt: 252.5 °C (760 mm Hg)
Melting Pt: 48…51 °C
Flash Pt: 120 °C
Storage Temperature: Ambient
MDL Number: MFCD01076323
CAS Number: 6833-47-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty