Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-4-Hydroxycinnamic acid 95%

Supplier: AMBEED, INC
Warning

A192873-5G A192873-25G A192873-5G A192873-1000G A192873-100G A192873-100MG A192873-10G A192873-500G
103847-100EA 6 USD InStock
103847-100 77084-990 77084-992 77260-524 77260-526 77260-528 77260-530 77260-532
trans-4-Hydroxycinnamic acid 95%
trans-4-Hydroxycinnamic acid
Precaución: for Research Use Only


Formula: C₉H₈O₃
Boiling Pt: 329.3 °C (760 mm Hg)
Melting Pt: 214 °C
Flash Pt: 167.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00004399
CAS Number: 501-98-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty