Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-4-Methoxycyclohexan-1-amine 95%

Supplier: AMBEED, INC
Warning

A154089-1G A154089-5G A154089-100MG A154089-250MG A154089-1G
103804-604EA 25 USD InStock
103804-604 103842-784 77028-288 77028-290 77318-480
trans-4-Methoxycyclohexan-1-amine 95%
trans-4-Methoxycyclohexan-1-amine
Precaución: for Research Use Only


Formula: C₇H₁₅NO
Storage Temperature: Ambient
MDL Number: MFCD20278175
CAS Number: 121588-79-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty