Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tri(2-furyl)phosphine 98%

Supplier: AMBEED, INC
Warning

A112931-1G A112931-5G A112931-100G A112931-250MG A112931-25G A112931-1G A112931-5G A112931-500G
103798-134EA 9 USD InStock
103798-134 103823-968 76781-222 76781-224 76781-226 77310-802 77310-804 77666-642
Tri(2-furyl)phosphine 98%
Tri(2-furyl)phosphine
Precaución: for Research Use Only


Formula: C₁₂H₉O₃P
Boiling Pt: 286.4 °C (760 mm Hg)
Melting Pt: 59…64 °C
Storage Temperature: Ambient
MDL Number: MFCD00151857
CAS Number: 5518-52-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty