Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethylstearylammonium chloride 98%

Supplier: AMBEED, INC
Warning

A909294-25G A909294-100G A909294-500G
103841-366EA 5 USD InStock
103841-366 76845-642 77007-052 77007-054 77007-056
Trimethylstearylammonium chloride 98%
Trimethylstearylammonium chloride
Precaución: for Research Use Only


Formula: C₂₁H₄₆ClN
MW: 348.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00050188
CAS Number: 112-03-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty