Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trimethylsulphoxonium iodide 98%

Supplier: AMBEED, INC
Warning

A108841-1000G A108841-100G A108841-25G A108841-500G
103808-006EA 118 USD InStock
103808-006 76780-978 76780-980 76780-982
Trimethylsulphoxonium iodide 98%
Trimethylsulphoxonium iodide
Precaución: for Research Use Only


Formula: C₃H₉IOS
MW: 220.07 g/mol
Melting Pt: 208…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00011899
CAS Number: 1774-47-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty