Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Triphenylsilanol 98%

Supplier: AMBEED, INC
Warning

A550917-100G A550917-10G A550917-500G A550917-25G
103839-822EA 9 USD InStock
103839-822 76790-248 76790-250 76790-252 76843-892
Triphenylsilanol 98%
Triphenylsilanol
Precaución: for Research Use Only


Formula: C₁₈H₁₆OSi
MW: 276.41 g/mol
Boiling Pt: 389 °C at [760 mmHg
Melting Pt: 150…153 °C
Flash Pt: 200 °C
Storage Temperature: Ambient
MDL Number: MFCD00002102
CAS Number: 791-31-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty