Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine 97%

Supplier: AMBEED, INC
Warning

A237566-5G A237566-250MG A237566-1G
103811-160EA 87 USD InStock
103811-160 103850-938 76874-354 77090-028
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine 97%
Tris[(1-benzyl-1H-1,2,3-triazol-4-yl)methyl]amine
Precaución: for Research Use Only


Formula: C₃₀H₃₀N₁₀
Melting Pt: 132…143 °C
Storage Temperature: Freezer
MDL Number: MFCD09265124
CAS Number: 510758-28-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty