Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tris(4-chlorophenyl)phosphine 97%

Supplier: AMBEED, INC
Warning

A358152-1G A358152-1G A358152-25G A358152-5G A358152-100G A358152-10G
103826-202EA 7 USD InStock
103826-202 103837-208 76840-368 76840-370 76840-372 77668-664 77872-543
Tris(4-chlorophenyl)phosphine 97%
Tris(4-chlorophenyl)phosphine
Precaución: for Research Use Only


Formula: C₁₈H₁₂Cl₃P
MW: 365.63 g/mol
Boiling Pt: 431
Melting Pt: 103
Flash Pt: 212.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00013639
CAS Number: 1159-54-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty