Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Hydroxy-3-methoxybenzyl alcohol 95%

Supplier: AMBEED, INC
Warning

A266033-25G A266033-100G A266033-100MG A266033-10G A266033-500G A266033-1000G
103852-674EA 5 USD InStock
103852-674 77092-372 77289-348 77289-350 77289-352 77289-354 77748-507
4-Hydroxy-3-methoxybenzyl alcohol 95%
4-Hydroxy-3-methoxybenzyl alcohol
Precaución: for Research Use Only


Formula: C₈H₁₀O₃
MW: 154.17 g/mol
Boiling Pt: 293
Melting Pt: 113
Flash Pt: 143.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00004659
CAS Number: 498-00-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty