Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Xipamide 97%

Supplier: AMBEED, INC
Warning

A200352-10MG A200352-50MG A200352-1G A200352-10MG A200352-50MG A200352-100MG A200352-1MG A200352-250MG
103792-376EA 15 USD InStock
103792-376 103799-474 103817-192 77085-878 77085-880 77152-126 77152-128 77152-130
Xipamide 97%
Xipamide
Precaución: for Research Use Only


Formula: C₁₅H₁₅ClN₂O₄S
MW: 354.81 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00865927
CAS Number: 14293-44-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty