Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Z-Asp(OMe)-OH 97%

Supplier: AMBEED, INC
Warning

A352933-1G A352933-25G A352933-5G A352933-100G A352933-10G
103826-080EA 6 USD InStock
103826-080 103837-080 76840-184 76840-186 76840-188 77003-274 77003-276 77003-278
Z-Asp(OMe)-OH 97%
Z-Asp(OMe)-OH
Precaución: for Research Use Only


Formula: C₁₃H₁₅NO₆
Boiling Pt: 503.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00133585
CAS Number: 3160-47-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty