Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cbz-D-Ser-OH 95%

Supplier: AMBEED, INC
Warning

A113078-1G A113078-100G A113078-10G A113078-250MG A113078-25G A113078-500G A113078-1G A113078-5G
103798-160EA 8 USD InStock
103798-160 103824-000 76810-034 76810-036 76810-038 76810-040 76810-042 77310-834 77310-836
Cbz-D-Ser-OH 95%
Cbz-D-Ser-OH
Precaución: for Research Use Only


Formula: C₁₁H₁₃NO₅
Melting Pt: 116…119 °C
Flash Pt: 248.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00063144
CAS Number: 6081-61-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty