Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

ZLN005 97%

Supplier: AMBEED, INC
Warning

A100765-1G A100765-10MG A100765-1MG A100765-250MG A100765-50MG A100765-5MG A100765-100MG
103795-616EA 226 USD InStock
103795-616 103821-312 77148-180 77148-182 77148-184 77148-186 77148-188 77684-926
ZLN005 97%
ZLN005
Precaución: for Research Use Only


Formula: C₁₇H₁₈N₂
Boiling Pt: 415.3±38.0 °C (760 mm Hg)
Flash Pt: 205.4 °C
Storage Temperature: Freezer
MDL Number: MFCD01560221
CAS Number: 49671-76-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty