Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid 95%

Supplier: AMBEED, INC
Warning

A193391-5G A193391-5G A193391-1000G A193391-100G A193391-500G
103847-148EA 9 USD InStock
103847-148 77274-546 77302-340 77748-231 77748-232 77748-233
(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid 95%
(S)-2-(Benzyloxycarbonylamino)-3-phenylpropanoic acid
Precaución: for Research Use Only


Formula: C₁₇H₁₇NO₄
Boiling Pt: 511.5 °C (760 mm Hg)
Melting Pt: 85…87 °C
Flash Pt: 263.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00020418
CAS Number: 1161-13-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty