Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Carbobenzoxy-L-threonine benzyl ester 98%

Supplier: AMBEED, INC
Warning

A176143-5G A176143-100G A176143-10G A176143-1G A176143-25G A176143-5G
103845-436EA 13 USD InStock
103845-436 76769-400 76769-402 76769-404 77301-428 77301-430
N-Carbobenzoxy-L-threonine benzyl ester 98%
N-Carbobenzoxy-L-threonine benzyl ester
Precaución: for Research Use Only


Formula: C₁₉H₂₁NO₅
MW: 343.38 g/mol
Boiling Pt: 536.8 °C (760 mm Hg)
Melting Pt: 78…82 °C
Flash Pt: 278.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077041
CAS Number: 16597-50-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty