Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

PermaSleeve® Polyolefin 16 to 8 Gauge Wire Marking Sleeves, 3" Core, Brady

Supplier: Brady Worldwide
PS-250-150-WT-S PS-250-2-WT-S 3PS-250-2-WT-S 3PS-250-2-YL 3PS-250-2-WT PS-250-2-WT PS-250-150-WT 3PS-250-2-WT-2 PS-250-2-WT-4 PS-250-150-WT-2 PS-250-2-WT-3 3PS-250-2-YL-2 3PS-250-2-WT-3 3PS-250-2-WT-S-2 PS-250-2-BK-S PS-250-2-YL-3 PS-250-2-WT-2 PS-250-2-YL
76461-174RL 622.3 USD InStock
76461-174 76461-184 76461-194 76461-202 76461-210 76461-226 76461-238 76461-244 76461-250 76461-256 76461-266 76461-270 76461-272 76461-280 76461-286 76461-360 76461-368 76251-027
PermaSleeve® Polyolefin 16 to 8 Gauge Wire Marking Sleeves, 3" Core, Brady
Labels
PermaSleeve® heat-shrink polyolefin (B-342) wire marking sleeves combine speed, ease of use, and permanence to create long-lasting, high-quality identification.

  • Flame-retardant
  • Heat-shrinkable
  • Wire diameter 0.25"
  • 3" core

The 3:1 heat shrinkable sleeve fits snugly around wires for maximum insulation protection and permanent identification. Fade-resistant and smudge-proof for superior aesthetic appearance. Cold resistant and flame-retardant with an operating temperature of –67 to +275 °F (–55 to +135 °C).

Sleeves are designed for use with BBP®11 Label Printer, BBP®12 Label Printer, BBP®72 Label Printer, Brady IP® Series Label Printer, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100 Industrial Label Printer, and THT 3" Core Printers.

Certificaciones: Meets SAE-AMS-DTL-23053/5C (Class 1), SAE-AS-81531, MIL-STD-202. RoHS compliant. UL recognized.

Información para pedidos: A ribbon is required to print this material. Use Part Number IP-R6600 for the BradyPrinter i5100 or BRADY IP® Series Label Printer; use R6600 for a different printer.

Order Now
 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty