Cuvette Standard O Optical Glass, Mettler-Toledo
Supplier: Mettler Toledo
30675053 30675054
76499-334EA 25.7 USD InStock
76499-334 76499-336
Cuvette Standard O Optical Glass, Mettler-Toledo
Cuvettes Spectrophotometer Cuvettes
The Standard Line O macro cuvette is made of fritted optical glass and comes in a matching pair. For measurements in the visible range with a wavelength between 320 nm – 2500 nm.
  • Reusable Glass Cuvettes
Accuracy by design
To ensure high accuracy and reliability of measurements, standard cuvettes come in matching pairs and are manufactured within tolerances of ± 0.05 mm.
Clear view
Standard line cuvettes provide good optical transparency > 80% and chemical stability.
A greener alternative
Glass cuvettes are designed for long-term use: This makes them an environmentally friendly choice that can also save money.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty