Helixyte™ Green dsDNA Quantifying Reagent 200X DMSO Solution, AAT BIOQUEST INC
Supplier: AAT BIOQUEST INC
17597 17598
76482-936EA 375.27 USD InStock
76482-936 76482-938
Helixyte™ Green dsDNA Quantifying Reagent 200X DMSO Solution, AAT BIOQUEST INC
Assays Nucleic Acid Assays
Helixyte™ Green is an excellent nucleic acid sensor that exhibits large fluorescence enhancement upon binding to dsDNA.
  • Same Spectral Properties To SYBR® Green
  • High Sensitivity
  • Useful For Nucleic Acid Labeling In Gels
Precaución: For Research Use Only
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty