Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Glu(Edans)-OH ≥95%

Supplier: AAT BIOQUEST INC
Warning

5009 5010 5011
76481-716EA 124.31 USD InStock
76481-716 76481-718 76481-720
Fmoc-Glu(Edans)-OH ≥95%
Fmoc-Glu(Edans)-OH
FMOC-Glu(EDANS)-OH is a building block for in-sequence peptide Glu labeling by EDANS.


Formula: C₃₂H₃₁N₃O₈S
MW: 617.673 g/mol
MDL Number: MFCD01076138
CAS Number: 193475-66-0
ADR: 5 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cf1f5807fc6bef9ade801ef3eb2ccdcdf630b58&vid=0cf1f5807fc6bef9ade801ef3eb2ccdcdf630b58&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty