Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-statine

Supplier: AAT BIOQUEST INC
Warning

5100 5101
76481-750EA 626.51 USD InStock
76481-750 76481-752
Boc-statine
Boc-statine
(3S,4S)-Statine is a novel amino acid of pepstatin, a low molecular weight inhibitor of acid proteases.


Formula: C₁₃H₂₅NO₅
MDL Number: MFCD00063180
CAS Number: 58521-49-6
ADR: 5,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a58e695b747ad87209f64935c6989917409e7482&vid=a58e695b747ad87209f64935c6989917409e7482&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty