Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2) trihydrate

Supplier: AAT BIOQUEST INC
Warning

11610 11612
76481-954EA 96.26 USD InStock
76481-954 76481-956
4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2) trihydrate
4-Methylumbelliferyl phosphate disodium salt (4-MUP-Na2) trihydrate
A fluorogenic substrate for phosphatases.


Formula: C₁₀H₇Na₂O₆P·3H₂O
MW: 354.16 g/mol
MDL Number: MFCD00037614
CAS Number: 22919-26-2
ADR: 5 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=502e62f616c54905c90d25bee4d6eb884ecb1d09&vid=502e62f616c54905c90d25bee4d6eb884ecb1d09&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty