Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Coelenterazine ≥95%

Supplier: AAT BIOQUEST INC
Warning

21150 21166
76483-302EA 96.26 USD InStock
76483-302 76483-306
Coelenterazine ≥95%
Coelenterazine
Calcium measurement is critical for numerous biological investigations.


Formula: C₂₆H₂₁N₃O₃
MW: 423.472 g/mol
MDL Number: MFCD00467176
CAS Number: 55779-48-1
ADR: 5 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5525bbecd54adea9b366d0615352ca0f9c566c48&vid=5525bbecd54adea9b366d0615352ca0f9c566c48&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty