Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Tenofovir

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS

46561-0010 46561-0050
TS46561-0010EA 89 USD InStock
TS46561-0010 TS46561-0050
Tenofovir
Tenofovir


Formula: C₉H₁₄N₅O₄P
MW: 287.22 g/mol
Melting Pt: 283°C
Storage Temperature: Freezer
MDL Number: MFCD00943794
CAS Number: 147127-20-6
Order Now
Specification Test Results
Appearance White to almost white powder
Infrared spectrum Conforms
Water ≤ 3%
HPLC ≥ 97.5%

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty