Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Crizotinib

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

46569-0010 46569-2500
TS46569-0010EA 570 USD InStock
TS46569-0010 TS46569-2500
Crizotinib
Crizotinib


Formula: C₂₁H₂₂Cl₂FN₅O
MW: 450.3 g/mol
Storage Temperature: Freezer
MDL Number: MFCD12407409
CAS Number: 877399-52-5
UN: 3077
ADR: 9 III
Order Now
Specification Test Results
Appearance White to beige powder
Infrared spectrum Conforms
HPLC ≥ 97.5%

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty