Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Mildronate dihydrate

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

46626-0010 466262500
TS46626-0010EA 198 USD InStock
TS46626-0010 TS466262500
Mildronate dihydrate
Mildronate dihydrate


Formula: C₆H₁₄N₂O₂·₂H₂O
MW: 182.22 g/mol
Melting Pt: 85°C…90°C
Storage Temperature: Refrigerator
MDL Number: MFCD00876389
CAS Number: 86426-17-7
Order Now
Specification Test Results
Appearance White to beige crystalline powder
Infrared spectrum Conforms
Titration with HClO4 97.5 to 102.5%
Water 18 to 22%

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty