Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Rotigotine

Supplier: THERMO FISHER SCIENTIFIC CHEMICALS
Warning

46654-1000 46654-5000
TS46654-1000EA 225 USD InStock
TS46654-1000 TS46654-5000
Rotigotine
Rotigotine


Formula: C₁₉H₂₅NOS
Storage Temperature: Ambient
MDL Number: MFCD00869590
CAS Number: 99755-59-6
Order Now
Specification Test Results
Appearance White to almost white powder
Infrared spectrum Conforms
HPLC ≥ 98%
Titration Total base 98 to 102%

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty