Virus Recombinant SARS-COV-2 RBD (FC TAG) (from Mammalian Cells)
Supplier: REVACC INC
W81X36-100UG W81X36-500UG
76515-444EA 386.23 USD InStock
76515-444 76515-446
Virus Recombinant SARS-COV-2 RBD (FC TAG) (from Mammalian Cells)
Proteins and Peptides
A recombinant form of the receptor binding domain (RBD with amino acids 333 to 529) of the spike (S) glycoprotein gene from severe acute respiratory syndrome-related coronavirus 2 (SARS-CoV-2), Wuhan-Hu-1 (GenBank: MN908947) was produced by human embryonic kidney HEK293F cells, followed by purification.
  • Protein function: Bind to its receptor
This product contains C-terminal human IgG Fc tag.
This product has been run on SDS-PAGE with reduced condition. The binding to Rabbit polyclonal mAb was tested in ELISAs.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty