Virus Recombinant Zika Virus E Protein Domain III (from Insect Cells)
Supplier: REVACC INC
X38F9-100UG X38F9-500UG
76515-460EA 386.23 USD InStock
76515-460 76515-462
Virus Recombinant Zika Virus E Protein Domain III (from Insect Cells)
Proteins and Peptides
A recombinant form of domain III of envelope (E) protein from Zika virus, (2015_Brazil strain, GenBank: ALU33341) was produced by insect S2 cells, followed by purification.
  • Protein function: Bind to its receptor
This product contains C-terminal 6 hexa-histidine tag.
This product has been run on SEC (superose 6) and SDS-PAGE with reduced condition. The binding to Flavivirus control mAb 4G2 (targets domain II of E protein), two published domain III-specific antibodies (ZV2 and ZV67) and one published Dengue cross-reactive antibody (EDE1-C8) was tested in ELISAs.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty