Brady® BradyJet™ J4000 GHS Labels, Brady
Supplier: Brady Worldwide
J40C-2250-7425J J40C-4000-7425J J40C-6000-7425J J40C-8000-7425J J40-260-7425J J40-262-7425J J40-265-7425J J40-321-7425J J40-268-7425J
76520-214RL 244.49 USD InStock
76520-214 76520-216 76520-218 76520-220 76520-256 76520-258 76520-260 76520-262 76520-264
Brady® BradyJet™ J4000 GHS Labels, Brady
Labels
This B-7425J polypropylene material is ideal for GHS and other full-color identification applications where resistance to harsh chemicals is required.
  • Recommended ribbon: J40-CMY
  • Chemical resistant, low temperature labels
  • Permanent acrylic adhesive type
  • Inkjet receptive polypropylene material
Matte white polypropylene with acrylic adhesive is inkjet printable. Low-cost, full-color GHS labeling option withstands chemical exposure. Service temperature of −112 to 158 °F (−80 to 70 °C). An ink cartridge is required to print this material. The recommended ribbon is J40-CMY.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty