CrossLab Bond Elut SAX, Agilent Technologies
Supplier: AGILENT TECHNOLOGIES, INC (CSD)
12102126 12162044B 12256021 12256029 14102017 14102044 14102079 14113043 14256013 14256029
AG12102126PK 197 USD InStock
AG12102126 AG12162044B AG12256021 AG12256029 AG14102017 AG14102044 AG14102079 AG14113043 AG14256013 AG14256029
CrossLab Bond Elut SAX, Agilent Technologies
Chromatography Columns
Bond Elut SAX is a strong anion-exchange sorbent well suited to compounds such as carboxylic acids.
  • Retains compounds that elute from weak anion-exchange sorbents
  • Selectivity can be user-modified for increased flexibility
  • Minimal nonpolar interactions
Bond Elut SAX is a strong anion-exchange sorbent ideally suited for the extraction of compounds, such as carboxylic acids, which may not retain effectively on weak anion-exchange sorbents.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty