Screen Quest™ Colorimetric ELISA cAMP Assay Kit, AAT Bioquest
Supplier: AAT BIOQUEST INC
36370 36371
76483-968EA 375.27 USD InStock
76483-968 76483-970
Screen Quest™ Colorimetric ELISA cAMP Assay Kit, AAT Bioquest
Assays ELISAs
Adenosine 3’, 5’ cyclic monophosphate (cAMP) is an important second messenger in intracellular signal transduction.
  • No acetylation step
  • Robust protocol
  • Sensitive substrate for easy quantification
Precaución: For research use only.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty