Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Coelenterazine n, Ultra Pure Grade calcium indicator

Supplier: AAT BIOQUEST INC
Warning

21155 21161
76483-324EA 124.31 USD InStock
76483-324 76483-326
Coelenterazine n, Ultra Pure Grade calcium indicator
Coelenterazine n
Calcium measurement is critical for numerous biological investigations.

  • Luminescent calcium probe
  • Brighter signal
  • Eliminates autofluorescence


Formula: C₃₀H₂₃N₃O₂
CAS Number: 123437-22-9
ADR: 5,II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcbb7a91491cf11330a46d87f8bce92b1ef581cc&vid=dcbb7a91491cf11330a46d87f8bce92b1ef581cc&prid=web&clid=SAPDAM https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c38b54ad0df42b13d07e39f97661f92957641f1&vid=3c38b54ad0df42b13d07e39f97661f92957641f1&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty