Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Trypan 0.1 M, Trypan Purple™ for dead cell labeling

Supplier: AAT BIOQUEST INC
Warning

2465 2466
76481-568EA 249.58 USD InStock
76481-568 76481-570
Trypan 0.1 M, Trypan Purple™ for dead cell labeling
Trypan
Trypan Purple™ is similar to Trypan Blue in cell permeability.

  • Minimal cytotoxicity
  • High purity


MW: 600 g/mol
ADR: 5 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ca120f065b1df940bad67da35567f543b5c04cc&vid=5ca120f065b1df940bad67da35567f543b5c04cc&prid=web&clid=SAPDAM
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty