Covipyte™ EN450 Peptide Substrate
Supplier: AAT BIOQUEST INC
13535 13536
76485-324EA 237.74 USD InStock
76485-324 76485-326
Covipyte™ EN450 Peptide Substrate
Proteins and Peptides
Covidyte™ EN450 is a peptide substrate containing 14 amino acid sequence (KTSAVLQSGFRKME) that can be cleaved by coronavirus proteases. The dark-FRET peptide contains Dabcyl (quencher) and Edans (donor) on the N-and C-terminals respectively where the fluorescence of Edans is effectively quenched by Dabcyl when the peptide is intact. When the peptide is hydrolyzed by coronavirus proteases, the Edans fragment generates significantly enhanced fluorescence since its fluorescence is no longer quenched by Dabcyl. The activity of coronavirus proteases can be effectively monitored by the fluorescence intensity of Edans. Covidyte™ EN450 is a convenient tool for screening inhibitors of coronavirus proteases.
  • MW 2584.98
Precaución: For research use only.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty