Covidyte™ IF670
Supplier: AAT BIOQUEST INC
13542 13543
76485-336EA 358.05 USD InStock
76485-336 76485-338
Covidyte™ IF670
Proteins and Peptides
Covidyte™ IF670 is a peptide substrate containing 12 amino acid sequence (VNSTLQSGLRKM) that can be cleaved by coronavirus proteases. The dark-FRET peptide contains Tide Quencher™ 5 (TQ5) as a quencher and iFluor® 670 as a fluorescent donor on the N-and C-terminals respectively where the fluorescence of iFluor® 670 is effectively quenched by TQ5 when the peptide is intact. When the peptide is hydrolyzed by coronavirus proteases, the iFluor® 670 fragment generates significantly enhanced fluorescence since its fluorescence is no longer quenched by TQ5. The activity of coronavirus proteases can be effectively monitored by the fluorescence intensity of iFluor® 670. Covidyte™ IF670 is a robust high throughput screening tool for searching inhibitors of coronavirus proteases. Comparing to the commonly used EDANS substrates (such as Covidyte™ ED450), the iFluor® 670 substrate has much stronger and longer fluorescence that is less interfered by colored compounds that often cause false positive hits.
  • MW 3067.69
Precaución: For research use only.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty