Cole-Parmer® Essentials Narrow-Mouth Plastic Bottles, HDPE, Antylia Scientific
Supplier: Argos Technologies
06035-10 06035-20 06035-30 06035-50 06035-60 06035-70
76534-092PK 19.53 USD InStock
76534-092 76534-104 76534-116 76534-140 76534-146 76534-148
Cole-Parmer® Essentials Narrow-Mouth Plastic Bottles, HDPE, Antylia Scientific
Bottles Plastic Bottles
Chemical-resistant bottles stand up to most corrosives.
  • Withstands freezer storage down to −100 °C
  • Will not leak with PP screw caps
  • Meets USP Class VI requirements
These all-purpose bottles are popularly used for packaging, shipping, and storage because of their reliable durability. Rigid, translucent, and chemical-resistant, they can be used with most corrosives. All have leakproof polypropylene screw caps. Bottles are not autoclavable.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty