Human Recombinant CD127/IL-7RA Protein (Fc Tag) (from HEK293 Cells)
Supplier: wmf consumer eletric GmbH
10975-H02H-100 10975-H02H-1
103886-674EA 300.98 USD InStock
103886-674 103887-598
Human Recombinant CD127/IL-7RA Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the human IL7R (P16871-1) (Met1-Gly236, with mutations I66T, V138I) was expressed with the Fc region of human IgG1 at the C-terminus.
Order Now
ORDER
SPECIFICATIONS
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty