Cynomolgus/Rhesus Recombinant Angiopoietin-2/ANG2 Protein (Fc Tag) (from HEK293 Cells)
Supplier: wmf consumer eletric GmbH
90026-C01H-100 90026-C01H-1
103585-092EA 300.98 USD InStock
103585-092 76536-596
Cynomolgus/Rhesus Recombinant Angiopoietin-2/ANG2 Protein (Fc Tag) (from HEK293 Cells)
Proteins and Peptides
A DNA sequence encoding the cynomolgus / rhesus ANGPT2 (XP_005562586.1/XP_001097949.1) (Lys274-Phe495) was expressed with the Fc region of human IgG1 at the N-terminus. Cynomolgus and rhesus ANGPT2 sequences are identical.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty